Learn More
Corning™ Bottle Top Dispensers (BTD) Cylinders
Dispenses reagent to avoid losing solution during normal transfer
Brand:  Corning™ 6858
 View more versions of this product
Product Code. 16872100
  • £158.25 / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
  • Comes with four adapters with different diameters for universal use with multiple bottle sizes
  • Telescopic suction tube can be adjusted smoothly to different bottle heights
  • Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
  • Every dispenser is individually tested and delivered with a certificate of performance
  • Uses:
    • For dispensing organic and inorganic liquids
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    100 mL
    Dispensing Organic and Inorganic Liquids
    Four adapters