missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Bottle Top Dispensers (BTD) Cylinders
Dispenses reagent to avoid losing solution during normal transfer
£94.69 - £152.58
Specifications
For Use With (Application) | Dispensing Organic and Inorganic Liquids |
---|---|
Includes | Four adapters |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
16822100
|
Corning™
6853 |
2.5 mL |
£94.69
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16832100
|
Corning™
6854 |
5 mL |
£94.69
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16842100
|
Corning™
6855 |
10 mL |
£102.06
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16852100
|
Corning™
6856 |
25 mL |
£121.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16862100
|
Corning™
6857 |
50 mL |
£126.27
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16872100
|
Corning™
6858 |
100 mL |
£152.58
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
Description
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Specifications
Dispensing Organic and Inorganic Liquids | |
Four adapters |