Learn More
Invitrogen™ Bovine IL-6 Uncoated ELISA Kit
ELISA
Brand: Invitrogen™ ESS0029
Description
The Bovine IL-6 Uncoated ELISA Kit contains pre-matched antibody pairs and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of bovine IL-6. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
Specifications
P26892 | |
<78.1 pg/mL | |
HRP | |
ELISA | |
No significant cross-reaction with recombinant bovine IL-1ß, IL-2, IL-4, IL-8, IFNg, or TNFa | |
Bovine | |
280826 | |
Coating Antibody, Lyophilized Recombinant Standard, Detection Antibody, Streptavidin-HRP, Substrate Solution, Ready to use Stop Solution, 0.16M Sulfuric Acid, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards | |
Absorbance at 450 nm minus absorbance (at 550 nm) | |
RUO | |
2°C to 8°C | |
24 hrs |
78.1 to 5000 pg/mL | |
Bovine IL-6 ELISA Reagent Kit | |
Colorimetric ELISA (TMB substrate) with Plates | |
Serum, Supernatant | |
5 x 96 microplate wells | |
Colorimetric Microplate Reader | |
IL6 | |
HRP | |
5 x 96 Tests | |
Serum 100 μL, Supernatant 100 μL | |
IL-6 | |
2 hrs 50 mins |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.