Learn More
Trajan™ BP1 GC Capillary Columns
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
Brand:  Trajan™ 054062
 View more versions of this product
Product Code. 12975531
  • £536.00 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
GC Column for Environmental Analysis
0.32 mm
0.25 μm
30 m