Learn More
Trajan™ BP1 GC Capillary Columns
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
£299.00 - £980.00
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
10422171
|
Trajan™
054053 |
0.15 mm | 1 μm | 25 m |
£494.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10107540
|
Trajan™
054071 |
0.32 mm | 1 μm | 25 m |
£565.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
16682602
|
Trajan™
054064 |
0.32 mm | 0.5 μm | 12 m |
£327.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
16692602
|
Trajan™
054066 |
0.32 mm | 0.5 μm | 50 m |
£935.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
12965531
|
Trajan™
054096 |
0.53 mm | 5 μm | 50 m |
£980.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
12653016
|
Trajan™
054070 |
0.32 mm | 1 μm | 12 m |
£332.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10045400
|
Trajan™
054047 |
0.22 mm | 0.25 μm | 25 m |
£505.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10229050
|
Trajan™
054048 |
0.22 mm | 0.25 μm | 50 m |
£865.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
16672602
|
Trajan™
054051 |
0.22 mm | 0.25 μm | 60 m |
£865.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
15238633
|
Trajan™
054056 |
0.25 mm | 1 μm | 30 m |
£494.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10310371
|
Trajan™
054059 |
0.32 mm | 0.25 μm | 25 m |
£535.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
15268633
|
Trajan™
054069 |
0.32 mm | 0.5 μm | 60 m |
£935.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10441031
|
Trajan™
054065 |
0.32 mm | 0.5 μm | 25 m |
£550.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
15278633
|
Trajan™
054073 |
0.32 mm | 3 μm | 30 m |
£530.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
15298633
|
Trajan™
054085 |
0.32 mm | 5 μm | 60 m |
£935.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10762372
|
Trajan™
054086 |
0.53 mm | 1 μm | 12 m |
£337.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
16602612
|
Trajan™
054087 |
0.53 mm | 1 μm | 25 m |
£585.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
15208713
|
Trajan™
054806 |
0.53 mm | 5 μm | 30 m |
£590.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
15218713
|
Trajan™
054807 |
0.53 mm | 5 μm | 60 m |
£980.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
15238713
|
Trajan™
054812 |
0.25 mm | 0.5 μm | 60 m |
£845.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10372160
|
Trajan™
054809 |
0.53 mm | 3 μm | 60 m |
£955.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
12796557
|
Trajan™
054808 |
0.53 mm | 3 μm | 30 m |
£590.00
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||
15258713
|
Trajan™
054820 |
0.25 mm | 0.5 μm | 30 m |
£505.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
12945531
|
Trajan™
054045 |
0.25 mm | 0.25 mm | 60 mm |
£865.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
10298760
|
Trajan™
054046 |
0.22 mm | 0.25 mm | 12 mm |
£299.00
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
10177250
|
Trajan™
054040 |
0.22 mm | 0.1 μm | 12 m |
£299.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
12975531
|
Trajan™
054062 |
0.32 mm | 0.25 μm | 30 m |
£530.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
12654945
|
Trajan™
054044 |
0.25 mm | 0.25 μm | 30 m |
£494.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
Description
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
- Classic cross-linked dimethyl polysiloxane technology
- Low bleed
- Non-polar
- Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines