Learn More
Tocris Bioscience™ BRACO 19 trihydrochloride
Telomerase inhibitor
Brand: Tocris Bioscience™ 5312/50
This item is not returnable.
View return policy
Description
BRACO 19 trihydrochloride is a telomerase inhibitor (IC50 = 115 nM). Inhibits expression of human telomerase reverse transcriptase (hTERT), induces cellular senescence and inhibits growth of uterine cancer cells in vitro. Inhibits growth of uterine tumor xenografts in mice.Specifications
N,N'-[9[[4-(Dimethylamino)phenyl]amino]-3,6-acridinediyl]bis-1-pyrrolidinepropanamide trihydrochloride | |
50 mg | |
C35H43N7O2·3HCl | |
Solid | |
Bioactive Small Molecule |
Telomerase inhibitor | |
Soluble to 50 mM in water and to 50 mM in DMSO | |
703.14 | |
White | |
94% |
For Research Use Only.