missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BRACO 19 trihydrochloride
Telomerase inhibitor
£146.86 - £571.26
Specifications
Chemical Name or Material | N,N'-[9[[4-(Dimethylamino)phenyl]amino]-3,6-acridinediyl]bis-1-pyrrolidinepropanamide trihydrochloride |
---|---|
Synonym | Telomerase inhibitor |
Solubility | Soluble to 50 mM in water and to 50 mM in DMSO |
Molecular Formula | C35H43N7O2·3HCl |
Formula Weight | 703.14 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753148
|
Tocris Bioscience™
5312/10 |
10 mg |
£146.86
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18708098
|
Tocris Bioscience™
5312/50 |
50 mg |
£571.26
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Telomerase inhibitor (IC50 = 115 nM). Inhibits expression of human telomerase reverse transcriptase (hTERT), induces cellular senescence and inhibits growth of uterine cancer cells in vitro. Inhibits growth of uterine tumor xenografts in mice.Specifications
N,N'-[9[[4-(Dimethylamino)phenyl]amino]-3,6-acridinediyl]bis-1-pyrrolidinepropanamide trihydrochloride | |
Soluble to 50 mM in water and to 50 mM in DMSO | |
703.14 | |
White | |
94% |
Telomerase inhibitor | |
C35H43N7O2·3HCl | |
Solid | |
Bioactive Small Molecule |
Safety and Handling
Recommended Storage : Store at -20°C
For Research Use Only.