Learn More
Brady™ Solvent and Chemical Resistant Polyester Labels with Ribbon for M21 Printers
Designed to meet the rigors of laboratory applications, Brady’s B-488 material excels at resisting chemicals, oils, alcohols, and heat. Due to its inherent properties, this polyester can also be used for asset tracking, datacom identification, barcode labeling, and more.
Brand:  Brady™ 110936
69.17 GBP valid until 2024-12-20
Use promo code "21731" to get your promotional price.
 View more versions of this product
Product Code. 11523904
  • £103.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
For use in extreme high temperatures
  • Designed to meet the rigors of laboratory applications, Brady’s B-488 material excels at resisting chemicals, oils, alcohols, and heat.
  • Due to its inherent properties, this polyester can also be used for asset tracking, datacom identification, barcode labeling, and more.
  • Engineered Performance — High-tack permanent adhesive ideal for flat surfaces exposed to a wide variety of liquids, chemicals, alcohols, oils, and will even survive outdoor applications.
  • Chemical Resistant — Resists exposure to mineral spirits, 10% sodium hydroxide solution, 10% sulfuric acid solution, oils, isopropyl alcohol, jet fuel, and more.
  • Certifications — UL Recognized to UL969, RoHS compliant.
Specifications
Specifications
Autoclavable
Surface Mount
Black, White
Cartridge (Label)
Thermal Transfer
Non-sterile
M210 Label Printer, M210-LAB Label Printer, M211 Label Printer, BMP™21 LAB Label Printer, BMP™21-PLUS Label Printer
1 Roll
-40°F to +320°F
3.7 mil
21 ft. x 0.75 in.
6.40 m
19.05 mm
AGA, CSA, UL
Permanent Acrylic, Permanent
Matte
Polyester
Label Cartridge
General identification, Laboratory identification: Vial, Well plate
1
Rectangle
-40°C to +160°C
0.094 mm
21 ft.
0.75 in.
6.4 cm