missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyBondz™ BMP71 Thermal Transfer Portable Printer Labels

Used to identify property from components to finished products

£45.00 - £49.15

Specifications

Certifications/Compliance UL Recognised; CSA Accepted; AGA Approved; Halogen Free (DIN VDE 0472/part 815)
Description BMP71 Label Printer Labels
Adhesive Type Permanent Acrylic
Color White
Finish Gloss
View More Specs

Products 5
Product Code Brand Size (Metric) Size (English) Price Quantity & Availability  
Product Code Brand Size (Metric) Size (English) Price Quantity & Availability  
11735703
View Documents
Brady™
115116
16.5 x 5.08 mm 0.65 x 0.2 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11705703
View Documents
Brady™
114850
12.7 x 6.99 mm 0.5 x 0.275 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11795673
View Documents
Brady™
115111
25.4 x 12.7 mm 1 x 0.5 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11755673
View Documents
Brady™
115110
25.4 x 9.5 mm 1 x 0.37 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11715693
View Documents
Brady™
114726
19 x 6.3 mm 0.75 x 0.25 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • White glossy polyester
  • Available in various sizes

Applications:

Asset Tracking, Barcoding, Component ID, Equipment ID, General Identification, General Purpose ID, Patch Panel - ID Port, Rating Plates, Through Hole - Top

Specifications

Specifications

UL Recognised; CSA Accepted; AGA Approved; Halogen Free (DIN VDE 0472/part 815)
Permanent Acrylic
Gloss
BradyBondz Label
BMP™71 Portable Thermal Transfer Printer
-70°C to +110°C
BMP71 Label Printer Labels
White
Polyester (B-423)
Asset Tracking, Barcoding, Bottles, Component ID, Conical Tube, Deep Well Plate, Equipment ID, General Purpose ID, Petri Dish, Rating Plates, Through Hole - Top
Rectangle
0.076 mm
Videos
SDS
Documents

Documents

Product Certifications