Learn More
Brady™ FreezerBondz™ Ultra Thin Cryogenic Polyester Labels with Vial Top for 1 in. Small Core Printers
Durable Freezerbondz™ Polyester (B-492) markers are designed for laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
Brand:  Brady™ 142078
178.65 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 15231808
  • £235.00 / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Printable labels for thermal transfer printers
  • Specialized — Aggressive adhesive B-492 FreezerBondz™ labels are ideal for curved, frozen surfaces like vials, centrifuge samples, tubes, and slides.
  • Durable — Labels withstand chemicals, abrasions, storage in liquid nitrogen, and a high/low temperature range of -320°F to +212°F (-196°C to +100°C).
  • Adhesive — Permanent acrylic adhesive for tight bonding onto smooth, curved surfaces, or wrapping around tubes and vials with a 1/8 in. overlap.
  • Certifications — RoHS compliant to 2011/65/EU (Pass 2015/863).
Specifications
Specifications
Not Autoclavable
Permanent Acrylic, Permanent
Matte
Printer Label
General identification, Laboratory identification
1500/Pk.
25.4 x 12.7 mm (W x H) with 11.18 mm Dia. vial top label
-196 to 120°C
25.40 mm
12.70 mm
2.54 cm
ASTM D 1000
White
Polyester
Non-sterile
Thermal printer
1 x 0.5 x 0.440 in. (W x H x dia.)
-320°F to +248°F
1 in.
0.5 in.
0.440 in.