Learn More
Brady™ Label Maker Vinyl Cartridges (B-595) for BMP51, BMP53, BMP41 Printers
Indoor/Outdoor grade vinyl conforms well to smooth or rough surfaces
Brand:  Brady™ 131583
Code : NEW
Additional Details : Weight : 0.07480kg
 View more versions of this product
Product Code. 15103359
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Durable and low-shrink with aggressive adhesive
  • Used on surfaces such as pipes, walls, doors, panels, shelving, bins and equipment
  • Offers resistance to oils, detergents, dirt, grime and many solvents
Specifications
Specifications
Black, Yellow
Label Maker Cartridge
1 Ea.
7.62 m x 25.4 mm
1 in.
7.62 m
Vinyl (B-595)
BMP™51,BMP™53,BMP™41 Printers
25 ft. x 1 in.
25 ft.
25.4 mm