Learn More
Brady™ Thermal Transfer Printable Nylon Cloth Labels
Thermal Transfer Printable Nylon Cloth Labels with high adhesion for use in autoclave, freezer or liquid nitrogen.
Brand:  Brady™ 116384
 View more versions of this product
Product Code. 30041427
  • £292.00 / Pack of 1500
Explore more special offers
Description
Description
  • White, matte finished labels suitable for use in laboratory identifications and vial labeling.
  • Low profile.
  • Recommended for use with Brady thermal transfer printer ribbon series 4900.
  • Compatible with color Brady series 6200, 6000, 4300, and 4500.
Specifications
Specifications
Autoclavable
THT-144-499-1.5-SC
Permanent Acrylic, Permanent
Matte
Nylon Cloth
Thermal Transfer
Non-sterile
BBP™11 Thermal Transfer Printer
3
-40 °C
Round
0.5 in. dia.
-40°F to +194°F
80 lb./in.
0.166 mm
0.5 in.
UL, CSA C22.2 No.0.15-95, RoHS 2002/95/EC, DIN VDE 0472 part 815
Thermal Transfer Printer Labels
White
Label
Single-side Printable
Labels
For wiremarking and general labeling applications
Roll of 1500 Labels
2 Years
1500 Pk.
0.5 in. (dia.)
1.3 cm dia.
-40°C to +90°C
0.0065 in.
15.88 mm
12.70 mm