Learn More
Brady™ TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels
TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels are ideal for cables, laboratory ID or general labeling where graphics need to be protected.
Brand:  Brady™ 062278
Code : 50
Additional Details : Weight : 0.04860kg
 View more versions of this product
Product Code. 10733183
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Labels for the TLS 2200™ and TLS-PC Link™ thermal transfer Portable Printers.
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
  • Matte white polyamide coated nylon cloth material (B-499) has a permanent acrylic adhesive with a service temperature of -40 to 193° F (-40 to 90° C) and recommended for indoor use.
  • Self-Laminating Polyester (B-461) has a permanent acrylic adhesive , good print smudge resistance, solvent resistance and high/low temperature performance.
Specifications
Specifications
RoHS
Acrylic, Permanent
Matte
Polyester
Non-sterile
BMP™61 Labeling System; BMP™71 Label Printer; TLS 2200™ Thermal Labeling System; TLS-PC LINK™ Thermal Labeling System
100/Roll
-196 °C to 110 °C
0.048 mm
2.625 in. H x 0.6 in. W
15.24 mm
66.675 mm
TLS 2200/TLS PC Link Labels
White
Label
TLS 2200™/TLS-PC Link™ Label
Centrifuge tube, Cryo vial, Eppendorf tube, Laboratory identification, PCR tube
1
Rectangle
0.0019 in.
69.85 mm
0.6 in.
2.62 in.