Learn More
Brady™ BradyJet™ Pre-Cut Polyester Labels
Brady™ BradyJet™ Pre-Cut Polyester Labels are for use with BradyJet™ J2000 and are ideal for a wide range of identification applications.
Brand:  Brady™ 150003
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 16120491
  • £295.00 / Each
Estimated Shipment: 06-09-2024
to see stock.
Add to basket
Description
Description
  • Constructed of indoor-use general purpose polyester
  • Features a specially-formulated topcoat for improved durability and color
  • Specially-formulated topcoat enhances colors and label appearance
  • Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
  • Topcoat protects print from smudging and harsh conditions
  • Inkjet-receptive labels for use with the BradyJet™ J2000 Color Label Printer
Specifications
Specifications
Acrylic
Matte
Polyester
BradyJet™ J2000
Rectangle
-40°C to +60°C
1.88 in. I.D.
White
Roll
Pipe & Valve Marking; Safety & Compliance ID; Visual Workplace Labeling; Warehouse Marking; Lockout Tagout Labeling; GHS & Chemical Labels; Arc Flash Labels
270/Roll
−40 to 140°F
4.33 x 2.95 in.