Learn More
Brady™ Bradylink™ Warning Post and Chain
This Bradylink™ Warning Post and Chain is made of colorful polystyrene and polyethylene and are a decorative alternative to metal posts.
Brand:  Brady™ 092121
 View more versions of this product
Product Code. 11761356
  • £689.00 / Pack of 6
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Posts are non-conductive so can be safely used in electrically hazardous areas.
  • Unlike metal posts, Bradylink™ Warning Posts are non-rusting.
  • Hollow base can be filled with sand for more stable application.
  • Large C-hook holds chain in place on large posts and can be used open or closed for greater security.
  • Bases and tops are included.
Specifications
Specifications
Warning Post and Chain
Yellow
Bradylink Warning Posts
Traffic Control
1041.4 mm
6 Pk.
BRADYLINK WARNING POST SOLID YELLOW-PAC
Polyethylene
3 in.
41 in.
Bases and tops