missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Bradylink™ Warning Post and Chain
This Bradylink™ Warning Post and Chain is made of colorful polystyrene and polyethylene and are a decorative alternative to metal posts.
£142.00 - £645.00
Specifications
Product Type | Warning Post and Chain |
---|---|
Material | Polyethylene |
Description | Bradylink Warning Posts |
Diameter (English) | 3 in. |
For Use With (Application) | Traffic Control |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
11721955
|
Brady™
092116 |
Black |
£142.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11741356
|
Brady™
092119 |
Black |
£645.00
Pack of 6 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11761356
|
Brady™
092121 |
Yellow |
£645.00
Pack of 6 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
Description
- Posts are non-conductive so can be safely used in electrically hazardous areas.
- Unlike metal posts, Bradylink™ Warning Posts are non-rusting.
- Hollow base can be filled with sand for more stable application.
- Large C-hook holds chain in place on large posts and can be used open or closed for greater security.
- Bases and tops are included.
Specifications
Warning Post and Chain | |
Bradylink Warning Posts | |
Traffic Control | |
1041.4 mm |
Polyethylene | |
3 in. | |
41 in. | |
Bases and tops |