missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Four-Clear Sided Disposable Polystyrene Cuvettes for Fluorescence
German manufactured premium cuvettes
Brand: BRAND™ 759030
15.64 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Additional Details : Weight : 0.27900kg
Description
- BRAND premium quality four sided clear disposable cuvettes for best results in fluorescence applications.
- Choose polystyrene for general purpose requirements in the visible range (340-800nm).
- Choose proprietary UV-plastic cuvettes with virtually no autoflourescence for best signal to noise ratio for assays in the 230-800nm range.
- 10mm lightpath to fit most fluorimeters.
Specifications
Fluorimeter Cuvette | |
2.5 to 4.5 mL | |
12.5mm | |
10 mm | |
Fluorometers | |
Fluorimetry |
BRAND four sided clear cuvette for fluorimetry. Polystyrene, package of 100 | |
Polystyrene | |
100 | |
45 mm | |
12.5 mm |