missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Four-Clear Sided Disposable Polystyrene Cuvettes for Fluorescence
German manufactured premium cuvettes
£18.94 - £75.60
Specifications
Type | Fluorimeter Cuvette |
---|---|
Capacity (Metric) | 2.5 to 4.5 mL |
Material | Polystyrene |
Length (Metric) | 12.5mm |
Path Length | 10 mm |
Description
- BRAND premium quality four sided clear disposable cuvettes for best results in fluorescence applications.
- Choose polystyrene for general purpose requirements in the visible range (340-800nm).
- Choose proprietary UV-plastic cuvettes with virtually no autoflourescence for best signal to noise ratio for assays in the 230-800nm range.
- 10mm lightpath to fit most fluorimeters.
Specifications
Fluorimeter Cuvette | |
Polystyrene | |
10 mm | |
12.5 mm |
2.5 to 4.5 mL | |
12.5mm | |
45 mm | |
Fluorimetry |