missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ 96-well Deep-well Plates
These deep-well plates are stackable for easy storage.
Brand: BRAND™ 701340
158.66 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- DNA-, DNase- and RNase-free
- Designed for a wide range of applications, such as High-Throughput Screening (HTS), tests requiring a mother plate, cell and tissue cultures, serial dilutions, reagent transfer and sample storage down to -80°C
- Deep-well plates are manufactured according to ANSI/SLAS 1 and 4 in.
- Choose PP plates for autoclavability and chemical resistance.
- Choose PS plates for transparency and surface properties.
- Deep Well Plates are ideal for sample storage, HTS assays requiring a motherplate, cell and tissue culture, immunological assays, and other applications.
Specifications
Standard | |
Clear | |
Polypropylene | |
96 | |
Round Bottom (U-well) | |
U-bottom |
8 x 12 | |
Deep-well Plate | |
1.2 mL | |
Non-sterile | |
Cell and tissue culture, sample storage, immunological assays and other applications |