missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ 96-well Deep-well Plates
These deep-well plates are stackable for easy storage.
Marca: BRAND™ 701348
Detalles adicionales : Weight : 5.50000kg
Descripción
- DNA-, DNase- and RNase-free
- Designed for a wide range of applications, such as High-Throughput Screening (HTS), tests requiring a mother plate, cell and tissue cultures, serial dilutions, reagent transfer and sample storage down to -80°C
- Deep-well plates are manufactured according to ANSI/SLAS 1 and 4 in.
- Choose PP plates for autoclavability and chemical resistance.
- Choose PS plates for transparency and surface properties.
- Deep Well Plates are ideal for sample storage, HTS assays requiring a motherplate, cell and tissue culture, immunological assays, and other applications.
Especificaciones
No Barcode | |
Standard | |
8 x 12 | |
Clear | |
Deep-well Plate | |
Polypropylene | |
96 | |
Square, Conical Bottom (V-well) | |
47 cm | |
18 cm |
ANSI/SLAS, Certified Life Science Quality, BIO-CERT™ PCR Quality | |
96-well Deep-well plate, PP, V-bottom, well shape square, suitable for KingFisherTM, 2,2 ml | |
Autoclavable | |
Single-use | |
KingFisher™ | |
2.2 mL | |
Non-sterile | |
Standard | |
47 cm | |
PCR Ready |