Learn More
BRAND™ BRANDplates™ pureGrade™ 96-Well Microplates
Constructed of polystyrene with nontreated surface. BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.
Brand:  BRAND™ 781600
93.36 GBP valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Code : 03
Additional Details : Weight : 4.26000kg
 View more versions of this product
Product Code. 10246473
  • £115.00 / Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
  • Polystyrene surface
  • Medium binding, nonsterile
  • Standard plate for most applications
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Strip plates available
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottoms available in F- shape
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
  • Suited for homogenous assays, screening, and storage.
Specifications
Specifications
Includes Barcode
Standard
8 x 12
Polystyrene
96
Non-treated
Yes
100/Cs.
ANSI, SLAS
Without Lid
Clear
330 μL
Non-sterile
Round Bottom (U-well)
Homogenous assays, screenings and storage
Yes