missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ Reagent Bottles with Stoppers
VITLAB polypropylene reagent bottles with polypropylene stoppers have good chemical resistance. Autoclavable at 121 C for 20 minutes.
Brand: BrandTech™ 100594
Additional Details : Weight : 1.67000kg
Description
- NS stoppers and good chemical resistance for long-term storage of liquids.
- Autoclavable at 121 C, according to DIN EN 285
Specifications
16.9 oz., 500mL | |
16.9 oz. | |
Narrow |
500 mL | |
Polypropylene | |
Polypropylene |