missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrandTech™ Reagent Bottles with Stoppers
VITLAB polypropylene reagent bottles with polypropylene stoppers have good chemical resistance. Autoclavable at 121 C for 20 minutes.
£2.65 - £65.60
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15782036
|
BrandTech™
100394 |
100 mL |
£44.95
Pack of 20 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
15792036
|
BrandTech™
100494 |
250 mL |
£65.60
Pack of 20 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
15702046
|
BrandTech™
100594 |
500 mL |
£47.85
Pack of 10 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
10396041
|
BrandTech™
100494 |
250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- NS stoppers and good chemical resistance for long-term storage of liquids.
- Autoclavable at 121 C, according to DIN EN 285
Specifications
Polypropylene | |
Polypropylene |