Learn More
BRAND™ Reagent Reservoirs
Constructed of heavy-gauge polypropylene for long-lasting durability. Reagent Reservoirs are autoclavable and suitable for all 8- and 12-channel pipetters. Non-sterile version includes lid.
Brand:  BRAND™ 703411
 View more versions of this product
Product Code. 10347392
  • £220.00 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Heavy-gauge polypropylene
  • Suitable for all 8- and 12-channel pipetters
  • Autoclavable
  • Non-sterile version includes lid
TRUSTED_SUSTAINABILITY
Specifications
Specifications
60 mL
Sterile
For use with multichannel pipets. Without lid.
Autoclavable
Polypropylene
Reagent Reservoir
Individually Wrapped