Learn More
Invitrogen™ BrightComp eBeads™ Compensation Bead Kits
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
Brand: Invitrogen™ A54740
Additional Details : Weight : 0.00600kg
Description
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
• Three levels of intensity, perfect for any samples no matter the level of expression• Near-identical, spectrally-matched dye to fluorescent protein
• Consistent, reliable performance
Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
Specifications
RFP BrightComp eBeads™ Compensation Bead Kit | |
Compensation Beads | |
Flow Cytometry | |
561 nm |