Learn More
Thermo Scientific™ Brij™-35 Surfact-Amps™ Detergent Solution
Stabilized Brij-35 detergent provided as 10% and 30% solutions for use in various protein methods including cell lysis and HPLC.
£166.00 - £880.00
Specifications
Description | Brij-35 Surfact-Amps Detergent Solution |
---|---|
Content And Storage | Store at Room Temperature |
Form | Liquid |
Product Line | Brij™, Surfact-Amps™ |
Reagent Type | Detergent Solution |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11841455
|
Thermo Scientific™
28316 |
Brij-35 Surfact-Amps Detergent Solution | 6 x 10 mL |
£166.00
Pack of 6 |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
13424269
|
Thermo Scientific™
85118 |
Brij-35 Surfact-Amps Detergent Solution | 500 mL |
£880.00
500mL |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
13414269
|
Thermo Scientific™
85117 |
Brij-35 Surfact-Amps Detergent Solution | 50 mL |
£169.00
50mL |
Estimated Shipment: 29-07-2024
Log in to see stock. |
||||
Description
Thermo Scientific™ Brij-35 Detergent Solutions are stabilized 10% and 30% solutions of Brij-35 detergent for use in various protein methods.
Additional Information and Online Ordering »
Brij-35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or as a surfactant in various HPLC applications. The detergent is offered in two forms: a Thermo Scientific Surfact-Amps™ Detergent Solution (high-purity, 10% solution) and a standard grade 30% solution.
Surfact-Amps Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neet detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules, ensuring their stability and eliminating the accumulation of peroxides and degradation products.
Highlights:
Brij-35 – a nonionic detergent for use in various protein methods
Accurate – 10% and 30% detergent solutions in ultrapure water
Easy-to-use – solutions are simple to dispense and dilute for use
Detergent Properties:
- Molecular Weight: 1225g
- Detergent Class: Nonionic
- Aggregation Number: 40
- Micelle Molecular Weight: 49,000g
- Critical Micelle Concentration (CMC): 0.09mM (0.011%, w/v)
- Cloud Point: >100°C
- Dialyzable: No
Surfact-Amps Detergent Solution (Part No. 28316):
Accurate – precise 10% detergent solution in ultrapure water
Easy-to-use – solution is simple to dispense and dilute for use
Exceptionally pure – less than 1.0μeq/ml peroxides and carbonyls
Stable – packed in glass ampules under inert nitrogen gas
Specifications
Brij-35 Surfact-Amps Detergent Solution | |
Liquid | |
Detergent Solution |
Store at Room Temperature | |
Brij™, Surfact-Amps™ |
For Research Use Only. Not for use in diagnostic procedures.