missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35, Thermo Scientific Chemicals
£24.62 - £236.18
Chemical Identifiers
CAS | 9002-92-0 |
---|---|
Molecular Formula | C14H30O2 |
Molecular Weight (g/mol) | 230.39 |
MDL Number | MFCD00043063 |
InChI Key | SFNALCNOMXIBKG-UHFFFAOYSA-N |
Synonym | Polyoxyethylene(23)lauryl ether |
IUPAC Name | 2-(dodecyloxy)ethan-1-ol |
SMILES | CCCCCCCCCCCCOCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10124810
|
Thermo Scientific Acros
228342500 |
250 g | HDPE plastic bottle |
£24.62
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10043100
|
Thermo Scientific Acros
228345000 |
500 g | HDPE plastic bottle |
£40.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10226430
|
Thermo Scientific Acros
228340010 |
1 kg | HDPE plastic bottle |
£69.95
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11956941
|
Thermo Scientific Acros
228340051 |
5 kg | Plastic drum |
£236.18
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9002-92-0 | |
230.39 | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
2-(dodecyloxy)ethan-1-ol |
C14H30O2 | |
MFCD00043063 | |
Polyoxyethylene(23)lauryl ether | |
CCCCCCCCCCCCOCCO |
Specifications
White | |
9002-92-0 | |
>100.0°C | |
>149°C | |
Authentic | |
C12H25(OCH2CH2)23OH | |
MFCD00043063 | |
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, propylene glycol, insoluble in mineral oil and vegetable oils | |
CCCCCCCCCCCCOCCO | |
230.39 | |
Brij™ 35 |
5mg KOH/g max. | |
Waxy Pellets | |
33.0°C to 40.0°C | |
40 to 60mg KOH/g | |
C14H30O2 | |
1% max. | |
Polyoxyethylene(23)lauryl ether | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
2-(dodecyloxy)ethan-1-ol | |
1199.56 |
Safety and Handling
Product Identifier
- Brij]TS 35
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brij™ L23 PHARMA-PA-(MV), previous name Brij™ 35, Thermo Scientific Chemicals