missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ CampyCount Agar

Product Code. p-6220030
Click to view available options
Format:
90mm Monoplate
Manufacturing Origin:
United Kingdom
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12719080

Brand: Thermo Scientific™ PO1185A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Accurately and efficiently identify and enumerate Campylobacter jejuni and C. coli from poultry samples on plate reader friendly, chromogenic plates.

Accurately, specifically and easily enumerate Campylobacter jejuni and C. coli from poultry and related samples using Thermo Scientific™ Brilliance™ CampyCount Agar. The transparent medium, on which Campylobacter produce distinct dark red colonies, makes identification and enumeration significantly easier than on traditional charcoal or blood-containing agar. The transparency of the medium also allows enumeration using plate readers for even greater efficiency.

Campylobacter is endemic in many poultry populations, with Campylobacter jejuni and Campylobacter coli being responsible for the majority of infections. As legislation moves towards enumerative limits on Campylobacter in poultry, food producers will put pressure on poultry companies to provide meat with low numbers of Campylobacter to reduce human infection.

Brilliance CampyCount Agar has been validated and approved by MicroVal (according to ISO 16140: 2003 standard) against the reference method ISO/TS 10272-2: 2006 for the selective enumeration of thermotolerant Campylobacter species, in particular C. jejuni and C. coli, in poultry products. This study included both the O.B.I.S. Campy kit and Oxoid™ Dryspot Campylobacter test as alternative confirmation methods to those described in the reference method ISO/TS 10272-2: 2006.
  • Easy-to-read chromogenic medium producing dark red colonies on a transparent background
  • Increase efficiency and accuracy - transparent medium allows enumeration on plate readers
  • Fast results - confirmed Campylobacter count in as little as 72 hours when used in conjunction with O.B.I.S. campy (ID0800) or Oxoid™ Campylobacter Test kit (DR0150)
  • Ready-to-use convenience of prepared media
Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 10°C
Description Brilliance CampyCount Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5g ± 1g
Packaging Type Wrapped in single cellulose-based film wrap
Quantity 10/Pk.

Disclaimer: Not intended for IVD use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.