Learn More
Thermo Scientific™ Brilliance™ CampyCount Agar
Accurately and efficiently identify and enumerate Campylobacter jejuni and C. coli from poultry samples on plate reader friendly, chromogenic plates.
Brand: Thermo Scientific™ PO1185A
26.19 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Accurately, specifically and easily enumerate Campylobacter jejuni and C. coli from poultry and related samples using Thermo Scientific™ Brilliance™ CampyCount Agar. The transparent medium, on which Campylobacter produce distinct dark red colonies, makes identification and enumeration significantly easier than on traditional charcoal or blood-containing agar. The transparency of the medium also allows enumeration using plate readers for even greater efficiency.
Campylobacter is endemic in many poultry populations, with Campylobacter jejuni and Campylobacter coli being responsible for the majority of infections. As legislation moves towards enumerative limits on Campylobacter in poultry, food producers will put pressure on poultry companies to provide meat with low numbers of Campylobacter to reduce human infection.Brilliance CampyCount Agar has been validated and approved by MicroVal (according to ISO 16140: 2003 standard) against the reference method ISO/TS 10272-2: 2006 for the selective enumeration of thermotolerant Campylobacter species, in particular C. jejuni and C. coli, in poultry products. This study included both the O.B.I.S. Campy kit and Oxoid™ Dryspot Campylobacter test as alternative confirmation methods to those described in the reference method ISO/TS 10272-2: 2006.
- Easy-to-read chromogenic medium producing dark red colonies on a transparent background
- Increase efficiency and accuracy - transparent medium allows enumeration on plate readers
- Fast results - confirmed Campylobacter count in as little as 72 hours when used in conjunction with O.B.I.S. campy (ID0800) or Oxoid™ Campylobacter Test kit (DR0150)
- Ready-to-use convenience of prepared media
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
2°C to 10°C | |
90mm Monoplate | |
Agar | |
Wrapped in single cellulose-based film wrap |
Brilliance CampyCount Agar | |
United Kingdom | |
19.5g ± 1g | |
10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer: Not intended for IVD use.