Learn More
Thermo Scientific™ Brilliance™ MRSA 2 Agar
Obtain fast and accurate results with a selective chromogenic medium for screening clinical samples for the presence of Methicillin Resistant Staphylococcus aureus (MRSA).
Brand: Thermo Scientific™ PO1210A
30.26 GBP valid until 2024-12-31
Use promo code "23263" to get your promotional price.
Code : 64
Additional Details : Weight : 0.30000kg
Description
Obtain results after 18 hours with no need for re-incubation of negatives using Thermo Scientific™ Brilliance™ MRSA 2 Agar. Rapidly eliminate negative results and target colonized patients for isolation and appropriate treatment as early as possible.
Brilliance MRSA 2 Agar has been enhanced over that of the original formulation in two ways. New inhibitory components in the medium inhibit the growth of more non-target organisms. In addition, those organisms that do grow are more easily distinguished from distinctive blue MRSA colonies, through inclusion of a novel pink counter-stain, further improving ease of interpretation.Brilliance MRSA 2 Agar has been refined to give larger MRSA colonies a more consistent color than the previous formulation. The improved recovery of MRSA, together with the increased selectivity, makes the new Brilliance MRSA 2 Agar an invaluable and efficient tool in the implementation of infection control.
- Brilliance MRSA 2 Agar can be inoculated from a screening swab taken from hospital patient or staff, from an isolated colony or from a liquid suspension.
- MRSA grows as blue colonies which are very easy to read against the light colored, opaque background.
- Plates are incubated at 37ºC, and provide high sensitivity and specificity, with results available in just 18 hours. This allows a rapid response, so enabling the patient to receive the most appropriate treatment as early as possible, reducing the risk of onward transmission of infection.
- Accuracy minimizes costs by helping to ensure that only those in need receive what can be costly treatment.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Pale buff, opaque | |
Brilliance MRSA 2 Agar | |
United Kingdom | |
19.5g ± 1.0g | |
10/Pk. |
2°C to 10°C | |
90mm Monoplate | |
Agar | |
Flow Wrapped - Single Layer |