missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coomassie Brilliant Blue G-250, Fisher BioReagents
£56.75 - £92.10
Chemical Identifiers
CAS | 6104-58-1 |
---|---|
Molecular Formula | C47H48N3NaO7S2 |
Molecular Weight (g/mol) | 854.025 |
InChI Key | RWVGQQGBQSJDQV-UHFFFAOYSA-M |
Synonym | Coomassie™ Blue G-250 |
PubChem CID | 6328534 |
IUPAC Name | sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate |
SMILES | CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+] |
Description
Brilliant Blue G-250 is a general protein stain.Chemical Identifiers
6104-58-1 | |
854.025 | |
Coomassie™ Blue G-250 | |
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate |
C47H48N3NaO7S2 | |
RWVGQQGBQSJDQV-UHFFFAOYSA-M | |
6328534 | |
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+] |
Specifications
6104-58-1 | |
C47H48N3NaO7S2 | |
Pass Test | |
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+] | |
854.025 | |
608 to 612nm | |
5% max. | |
Blue | |
Brilliant Blue G-250 |
600g(-1) cm(-1) min. | |
Coomassie™ Blue G-250 | |
RWVGQQGBQSJDQV-UHFFFAOYSA-M | |
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate | |
6328534 | |
854.02 | |
42655 | |
Solid |
Safety and Handling
Recommended Storage : RT
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title