Learn More
Mettler Toledo™ Bromide Ion Selective Electrode
Brand:  Mettler Toledo™ 51340300
995.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10159750
  • £1030.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Specifications
Solid State Membrane
0°C to 80°C
2 to 13
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Mettler Toledo™ Bromide Ion Selective Electrode > Membrane: Solid State