Learn More
Bromine, 1M solution in acetic acid, Thermo Scientific Chemicals
£59.50 - £309.00
Specifications
Chemical Name or Material | Bromine |
---|---|
Color | Red |
Name Note | 1M solution in acetic acid |
CAS | 7726-95-6, 64-19-7 |
Molecular Formula | Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10530364
|
Thermo Scientific Acros
388401000 |
100 mL | AcroSeal™ Glass bottle |
£59.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10445465
|
Thermo Scientific Acros
388408000 |
800 mL | AcroSeal™ Glass Bottle |
£309.00
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Bromine | |
1M solution in acetic acid | |
Br | |
MFCD00010896 | |
3.111 | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
bromide | |
24408 | |
159.82 | |
Liquid |
Red | |
7726-95-6, 64-19-7 | |
Br2 | |
0.90 to 1.10M | |
15, 1401 | |
Solubility in water: soluble. | |
[Br-] | |
79.91 | |
CHEBI:29224 | |
40°C |
Safety and Handling
GHS P Statement Fatal if inhaled. Causes severe skin burns and eye damage. Very toxic to aquatic life. Flammable liquid and vapor.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid release to the environment.
GHS Signal Word: Danger
- Bromine, 1M solution in acetic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only