missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromine, 99.6%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent for analysis

Specifications
Specifications
Boiling Point | 58.7°C |
Melting Point | -7.2°C |
Quantity | 1 L |
Assay Percent Range | 99.5% min. (Iodometry) |
Linear Formula | Br2 |
Fieser | 03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47 |
Merck Index | 15, 1401 |
Solubility Information | Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2 |
IUPAC Name | bromide |
Formula Weight | 159.82 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468735 | Certificate of Analysis | 31/01/2025 | 196662500,196660250,196660010 |
A0468736 | Certificate of Analysis | 19/12/2024 | 196662500,196660250,196660010 |
A0466776 | Certificate of Analysis | 11/10/2024 | 196662500,196660250,196660010 |
A0465109 | Certificate of Analysis | 01/08/2024 | 196662500,196660250,196660010 |
A0465108 | Certificate of Analysis | 01/08/2024 | 196662500,196660250,196660010 |
Safety and Handling



Product Identifier
- Bromine
- Danger
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P273-Avoid release to the environment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bromine, 99.6%, for analysis, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction