Learn More
Bromine, 99+%, extra pure, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7726-95-6 |
---|---|
Molecular Formula | Br |
Molecular Weight (g/mol) | 79.91 |
MDL Number | MFCD00010896 |
InChI Key | CPELXLSAUQHCOX-UHFFFAOYSA-M |
Synonym | bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish |
PubChem CID | 24408 |
ChEBI | CHEBI:29224 |
SMILES | [Br-] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7726-95-6 | |
79.91 | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
24408 | |
[Br-] |
Br | |
MFCD00010896 | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
CHEBI:29224 |
Specifications
7726-95-6 | |
Fuming Liquid | |
Br | |
MFCD00010896 | |
15, 1401 | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
bromide | |
24408 | |
159.82 | |
Extra Pure | |
Bromine |
Red to Brown | |
99+% | |
Br2 | |
03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47 | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
[Br-] | |
79.91 | |
CHEBI:29224 | |
≥99.0% | |
3.1110g/mL |
Safety and Handling
GHS P Statement Causes severe skin burns and eye damage. Fatal if inhaled. Very toxic to aquatic life.
GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Avoid release to the environment.
Danger
- Bromine
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only