missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Bromophenol Blue Product Code.: 11488387

Thermo Scientific Chemicals Bromophenol Blue

Product Code. 11488387
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11488387

Brand: Thermo Scientific Chemicals A18469.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is employed as an electrophoresis tracking dye and also finds its uses as an industrial dye, a laboratory acid-base indicator and a biological stain to detect proteins and nucleic acids (absorbance at 610 nm), particularly when staining living tissues. In gel electrophoresis it can be used as a color marker and as a biological stain it can be used to stain proteins and nucleic acids. Bromophenol blue is an intermediate and is used as an acid-based indicator in the pH range of 3-4.6 where the color changes from yellow to blue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an electrophoresis tracking dye and also finds its uses as an industrial dye, a laboratory acid-base indicator and a biological stain to detect proteins and nucleic acids (absorbance at 610 nm), particularly when staining living tissues. In gel electrophoresis it can be used as a color marker and as a biological stain it can be used to stain proteins and nucleic acids. Bromophenol blue is an intermediate and is used as an acid-based indicator in the pH range of 3-4.6 where the color changes from yellow to blue.

Solubility
Freely soluble in NaOH. More soluble in methyl and ethyl alcohols ,benzene, and in acetic acid. Slightly soluble in water (0.4g/100g) at 20°C. Soluble in methanol (10 mg/mL.).

Notes
Transition interval: pH 3.0 (yellow) to pH 4.6 (blue)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
Synonym Bromphenol Blue
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point ∼270°C (decomposition)
Beilstein 61698
Merck Index 14,1444
Solubility Information Freely soluble in NaOH. More soluble in methyl and ethyl alcohols ,benzene,and in acetic acid. Slightly soluble in water (0.4g/100g) at 20°C. Soluble in methanol (10mg/mL.).
Formula Weight 669.98
Odor Odorless
Quantity 10 g
Chemical Name or Material Bromophenol Blue

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251113 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA18469.18,A18469.09
Lot Number10243922 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA18469.18,A18469.09
Lot Number10238987 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA18469.18,A18469.09
Lot Number10231827 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18469.18,A18469.09
Lot NumberL13J26 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18469.18,A18469.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Bromophenol Blue >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.