missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromophenol Blue, ACS

£26.93 - £94.27

Chemical Identifiers

CAS 115-39-9
Molecular Formula C19H10Br4O5S
Molecular Weight (g/mol) 669.96
MDL Number MFCD00005875
InChI Key UDSAIICHUKSCKT-UHFFFAOYSA-N
PubChem CID 8272
ChEBI CHEBI:59424
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,lambda {6}-benzoxathiol-3-yl]phenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
View More Specs

Produits 3
Code produit Marque Quantity Prix Quantité et disponibilité  
Code produit Marque Quantity Prix Quantité et disponibilité  
11362888
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
032641.03
1 g
£26.93
1g
In Stock
to see stock.
Ajouter au panier
 
11352888
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
032641.06
5 g
£31.46
5g
In Stock
to see stock.
Ajouter au panier
 
11342888
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
032641.14
25 g
£94.27
25g
In Stock
to see stock.
Ajouter au panier
 
Description

Description

Bromophenol blue is used as a tracking dye in electrophoresis, as a color marker in gel electrophoresis and polyacrylamide gel electrophoresis. As a biological stain, it plays an important role to stain proteins and nucleic acids. It acts an intermediate and an acid-base indicator in the pH range 3.0 to 4.6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécification

Identifiants chimiques

115-39-9
669.96
UDSAIICHUKSCKT-UHFFFAOYSA-N
CHEBI:59424
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
C19H10Br4O5S
MFCD00005875
8272
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,lambda {6}-benzoxathiol-3-yl]phenol

Spécification

270°C (decomposition)
C19H10Br4O5S
61698
Freely soluble in NaOH; Soluble in sodium hydroxide,alcohol,benzene,and acetic acid; Slightly soluble in water.
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Br)C4=CC(=C(C(=C4)Br)O)Br
669.96
CHEBI:59424
ACS
Bromophenol Blue
115-39-9
MFCD00005875
141444
UDSAIICHUKSCKT-UHFFFAOYSA-N
2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,lambda {6}-benzoxathiol-3-yl]phenol
8272
669.98
Crystalline
Vidéos
Sécurité et traitement

Sécurité et traitement

H500

missing translation for 'einecsNumber' : 204-086-2

missing translation for 'rtecsNumber' : SJ7453000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Product Certifications

RUO – Research Use Only