Learn More
Thermo Scientific Chemicals Bromophenol Blue, ACS
Description
Bromophenol blue is used as a tracking dye in electrophoresis, as a color marker in gel electrophoresis and polyacrylamide gel electrophoresis. As a biological stain, it plays an important role to stain proteins and nucleic acids. It acts an intermediate and an acid-base indicator in the pH range 3.0 to 4.6.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 270°C (decomposition) |
Beilstein | 61698 |
Merck Index | 141444 |
Solubility Information | Freely soluble in NaOH; Soluble in sodium hydroxide,alcohol,benzene,and acetic acid; Slightly soluble in water. |
Formula Weight | 669.98 |
Grade | ACS |
Odor | Odorless |
Quantity | 5 g |
Physical Form | Crystalline |
Chemical Name or Material | Bromophenol Blue |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P17L042 | Certificate of Analysis | 19/03/2025 | 032641.06,032641.03,032641.14 |
N07K036 | Certificate of Analysis | 13/02/2024 | 032641.06,032641.03,032641.14 |
W24G026 | Certificate of Analysis | 05/12/2023 | 032641.06,032641.03,032641.14 |
W22J041 | Certificate of Analysis | 06/10/2023 | 032641.06,032641.03,032641.14 |
M10J009 | Certificate of Analysis | 13/01/2023 | 032641.06,032921.DR,032921.DM,032641.03,032641.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.