missing translation for 'onlineSavingsMsg'
Learn More

Bromophenol Blue, Pure, Solution 0.04%, Indicator Grade, Fisher Chemical™

Brand:  Fisher Chemical B/4630L/05


 View more versions of this product

Product Code. 10660692

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5875
Blue
Liquid
Amber glass bottle
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bromophenol blue solution 0.04% in methylated spirits

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Acetone, Methyl alcohol,Bromphenol blue

SDS
Documents

Documents

Certificates
Special Offers

Special Offers