missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromophenol Blue, Pure, Solution 0.04%, Indicator Grade, Fisher Chemical™
Specifications
MDL Number | 5875 |
---|---|
Packaging | Amber glass bottle |
Color | Blue |
Physical Form | Liquid |
Description
Specifications
5875 | |
Amber glass bottle | |
Blue | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Bromophenol blue solution 0.04% in methylated spirits
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Ethyl alcohol, Acetone, Methyl alcohol,Bromphenol blue