missing translation for 'onlineSavingsMsg'
Learn More
Bromotrimethylsilane, 97%, stab. with copper powder or silver wire, Thermo Scientific Chemicals Product Code.: 11488236

Bromotrimethylsilane, 97%, stab. with copper powder or silver wire, Thermo Scientific Chemicals

Product Code. 11488236
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11488236

Brand: Thermo Scientific Alfa Aesar A15334.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromotrimethylsilane is used as a mild and selective reagent for cleavage of lactones, epoxides, acetals, phosphonate esters and certain ethers. Further, it is used as a brominating agent and an effective reagent for the formation of silyl enol ethers.

Solubility
Miscible with carbon tetrachloride, chloroform, dichloroethane, acetonitrile, toluene, dichloromethane and hexane.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids, water and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2857-97-8
Molecular Formula C3H9BrSi
Molecular Weight (g/mol) 153.094
MDL Number MFCD00000048
InChI Key IYYIVELXUANFED-UHFFFAOYSA-N
Synonym trimethylsilyl bromide, trimethylbromosilane, silane, bromotrimethyl, trimethylsilicon bromide, tmsbr, trimethylsilylbromide, bromotrimethyl silane, tmbs, bromo trimethyl silane, tms bromideShow MoreShow Less
PubChem CID 76113
IUPAC Name bromo(trimethyl)silane
SMILES C[Si](C)(C)Br

Specifications

Boiling Point 78°C to 80°C
Melting Point -44°C
Quantity 10 g
Linear Formula (CH3)3SiBr
UN Number UN2924
Beilstein 1731135
Flash Point −3°C (26°F)
Solubility Information Miscible with carbon tetrachloride,chloroform,dichloroethane,acetonitrile,toluene,dichloromethane and hexane.
Formula Weight 153.09
Percent Purity 97%
Odor Acrid
Refractive Index 1.425
Sensitivity Moisture sensitive
Density 1.183
Chemical Name or Material Bromotrimethylsilane, stabilized with copper powder or silver wire
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253719 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog NumberA15334.18,A15334.09,A15334.30
Lot Number10235161 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberA15334.18,A15334.09,A15334.30
Lot Number10243101 Certificate TypeCertificate of Analysis Date24/08/2022 Catalog NumberA15334.18,A15334.09,A15334.30
Lot Number10187715 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15334.18,A15334.09,A15334.30
Lot Number10131200 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15334.18,A15334.09,A15334.30
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromotrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Bromotrimethylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromotrimethylsilane, 97%, stab. with copper powder or silver wire, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.