Learn More
Bronze powder, -100 mesh, 99.0% (metals basis), Thermo Scientific Chemicals
£41.31 - £114.57
Specifications
Chemical Name or Material | Bronze |
---|---|
Appearance | Bronze colored powder |
Assay Percent Range | 99% (Metals basis) |
MDL Number | MFCD00198187 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389249
|
Thermo Scientific Alfa Aesar
088359.30 |
250 g |
£41.31
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399249
|
Thermo Scientific Alfa Aesar
088359.A1 |
1 kg |
£114.57
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bronze powder suitable for a range of applications including resin-casting, decorative coatings and powder metallurgy. Also used for hydraulic seals, surface engineering, decorative, welding and brazing, diamond tooling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bronze | |
99% (Metals basis) | |
Insoluble in water. | |
Odorless |
Bronze colored powder | |
MFCD00198187 | |
-100 Mesh | |
Powder |
Safety and Handling
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.