Learn More
Bronze powder, -100 mesh, 99.0% (metals basis), Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 088359.30
41.10 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11389249
  • £41.31 / 250g
Explore more special offers
Description
Description
Bronze powder suitable for a range of applications including resin-casting, decorative coatings and powder metallurgy. Also used for hydraulic seals, surface engineering, decorative, welding and brazing, diamond tooling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bronze
99% (Metals basis)
250 g
-100 Mesh
Powder
Bronze colored powder
MFCD00198187
Insoluble in water.
Odorless
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bronze powder, -100 mesh, 99.0% (metals basis), Thermo Scientific Chemicals > 250g