missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PolarScreen™ ER Alpha Competitor Assay, Green

Brand:  Thermo Scientific™ A15883

 View more versions of this product

Product Code. 15697848

  • £1530.00 / Pack of 800

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • The PolarScreen™ ER (Estrogen Receptor) Alpha Competitor Assay Kit, Green, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential ER alpha ligands. The kit uses insect cell-expressed, full-length, native (untagged) human estrogen receptor alpha (available separately, Cat. No. A15674) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone ES2 Green) in a homogenous, mix-and-read assay format.
  • This kit produces a green color read-out; a red kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation.
  • Complete & ready-to-use out of the box - all reagents included, just add your test compounds
  • High throughput compatible - 800 x 20 μL fluorescence polarization assays in a 384-well plate
  • Convenient - available in bulk sizes for larger screening needs
  • Includes:
    • 2 tubes full-length ER Alpha Recombinant Protein (store at -68 to -85°C)
    • 1 tube of Fluormone ES2 Green Tracer (store at -5 to -30°C)
    • 1 bottle of ES2 Screening Buffer (store at -5 to -30°C)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:

• 2 tubes full-length ER Alpha Recombinant Protein (store at -68 to -85°C)
• 1 tube of Fluormone ES2 Green Tracer (store at -5 to -30°C)
• 1 bottle of ES2 Screening Buffer (store at -5 to -30°C)
Fluorescent Imager
800 Assays
Fluorescence
384-well Plate
ER Alpha Competitor Assay
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2878582 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog NumberA15883
Lot Number2895003 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog NumberA15883
Lot Number2795003 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog NumberA15883
Lot Number2717017 Certificate TypeCertificate of Analysis Date31/05/2023 Catalog NumberA15883
Lot Number2669854 Certificate TypeCertificate of Analysis Date24/03/2023 Catalog NumberA15883
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ PolarScreen™ ER Alpha Competitor Assay, Green > 800 assays

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.