Learn More
Steris™ Browne™ Control™ Autoclave Indicator Valves
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
Brand: Steris™
Code : 33
Additional Details : Weight : 0.12420kg
Description
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
- The use of TST Control™ emulating Indicators is recommended for monitoring porous load steam sterilization processes
- Various colors
- Glass tubes
Specifications
Yellow Spot | |
Autoclave | |
134°C | |
Yes | |
Steam, Dry Heat |
For monitoring porous load steam sterilization processes | |
Browne Control Tube | |
100 Pk. | |
3 min. |