Learn More
Steris™ Browne™ Control™ Autoclave Indicator Valves
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
Brand: Steris™ 7303AB
Description
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
- The use of TST Control™ emulating Indicators is recommended for monitoring porous load steam sterilization processes
- Various colors
- Glass tubes
Specifications
Green Spot | |
Autoclave | |
160°C | |
Yes | |
Steam, Dry Heat |
For monitoring porous load steam sterilization processes | |
Browne Control Tube | |
100 Pk. | |
60 min. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Steris™ Browne™ Control™ Autoclave Indicator Valves > Sterilization temperature: 160deg.C; Color: green spot
Spot an opportunity for improvement?Share a Content Correction