Learn More
Steris™ Browne™ Control™ Autoclave Indicator Valves
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
Brand: Steris™ 7305AB
Description
Test the efficacy of an autoclave using Browne™ Control™ Autoclave Indicator Valves. These tubes contain heat-sensitive dyes that change color after sufficient time at the autoclave's desired temperature.
- The use of TST Control™ emulating Indicators is recommended for monitoring porous load steam sterilization processes
- Various colors
- Glass tubes
Specifications
White Spot | |
Autoclave | |
100 Pk. | |
Steam, Dry Heat |
For monitoring porous load steam sterilization processes | |
Browne Control Tube | |
Yes |
Product Suggestions
Customers who viewed this item also viewed