Learn More
BTK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21581-1-AP-20UL
 View more versions of this product
Product Code. 16851185
  • £135.00 / 20µL
Explore more special offers
Description
Description
The protein encoded by this gene plays a crucial role in B-cell development. Mutations in this gene cause X-linked agammaglobulinemia type 1, which is an immunodeficiency characterized by the failure to produce mature B lymphocytes, and associated with a failure of Ig heavy chain rearrangement.
Specifications
Specifications
BTK
Polyclonal
Unconjugated
BTK
AGMX1, AT, ATK, B cell progenitor kinase, BPK, Bruton tyrosine kinase, BTK, IMD1, PSCTK1, Tyrosine protein kinase BTK, XLA
Rabbit
Antigen Affinity Chromatography
RUO
12229, 367901, 695
-20°C
Liquid
Western Blot, Immunoprecipitation
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35991, Q06187
Btk
BTK Fusion Protein Ag16133
20 μL
Primary
Human, Rat, Mouse