missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | BTK |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16851185
|
Proteintech
21581-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16841185
|
Proteintech
21581-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene plays a crucial role in B-cell development. Mutations in this gene cause X-linked agammaglobulinemia type 1, which is an immunodeficiency characterized by the failure to produce mature B lymphocytes, and associated with a failure of Ig heavy chain rearrangement.Specifications
BTK | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P35991, Q06187 | |
12229, 367901, 695 | |
BTK Fusion Protein Ag16133 | |
Primary | |
-20°C | |
BTK |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AGMX1, AT, ATK, B cell progenitor kinase, BPK, Bruton tyrosine kinase, BTK, IMD1, PSCTK1, Tyrosine protein kinase BTK, XLA | |
Btk | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BTK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech