missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Tweezertrode™ Electrode Kit

Tweezertrode™ Electrode Kit features reusable, tweezer-style, in vivo electrodes suitable for drug or gene delivery in animals.

Brand:  BTX™ 450165

 View more versions of this product

Product Code. 15487280

  • £1760.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel or platinum at the tip
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned
  • Kits are supplied with BTX524 coaxial cable for connection to electroporation generators
Specifications

Specifications

In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery.
Stainless Steel
Electrode Kit
Stainless Tweezertrode Kit, 7 mm
7 mm
Cable
0.47 in.
12 mm
1 Ea.
<0.04 in. to >0.79 in.
11.5 cm
Banana Cables
ECM 830, Gemini X2, ECM 2001+, Legacy ECM 2001 Generator
BTX Electroporation
1000 V
0.27 in.
Circular
0.27 in.
7 mm
10 μsec. to 99 msec. in PBS
20 to 80%
<1 mm to >2 cm
Non-sterile
Square Wave
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Intended for Research use only. Not for use in diagnostic or clinical procedures.